2-Nitroterephthalsäure-1-methylester, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL515000-5GEA 28.9 EUR
SIAL515000-5G
2-Nitroterephthalsäure-1-methylester, Sigma-Aldrich®
2-Nitroterephthalsäure-1-methylester
Formel: C₉H₇NO₆
Molekulargewicht: 225,17
MDL Number: MFCD00024510
CAS-Nummer: 35092-89-8
EINECS: 252-360-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR