2-(2,4-Dichlorphenoxy)propionsäurehydrazid ≥98%

Lieferant: Thermo Scientific
Achtung

A16817.03EA 17.4 EUR
A16817.03 A16817.06 A16817.14
2-(2,4-Dichlorphenoxy)propionsäurehydrazid ≥98%
2-(2,4-Dichlorphenoxy)propionsäurehydrazid
Formel: C₉H₁₀Cl₂N₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00204152
CAS-Nummer: 15253-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR