2-(3-Chlorphenoxy)propionsäure
Lieferant: EHRENSTORFER GMBH
EHERC11688000EA
99.2
EUR
EHERC11688000
2-(3-Chlorphenoxy)propionsäure
2-(3-Chlorphenoxy)propionsäure
Formel:
C₉H₉ClO₃ |
MDL Number:
MFCD00009723 CAS-Nummer: 101-10-0 EINECS: 202-915-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...