2-(3-Chlorphenoxy)propionsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL233013-50GEA
87.4
EUR
SIAL233013-50G
2-(3-Chlorphenoxy)propionsäure, Sigma-Aldrich®
2-(3-Chlorphenoxy)propionsäure
Formel:
C₉H₉ClO₃ |
MDL Number:
MFCD00009723 CAS-Nummer: 101-10-0 EINECS: 202-915-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...