Tributyl(pentafluorethyl)stannan, Sigma-Aldrich®

Lieferant: Merck
Gefahr

SIAL711063-1GEA 98.2 EUR
SIAL711063-1G
Tributyl(pentafluorethyl)stannan, Sigma-Aldrich®
Tributyl(pentafluorethyl)stannan
Formel: C₁₄H₂₇F₅Sn
MDL Number: MFCD04116432
CAS-Nummer: 1426-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR