3-(4-Fluorphenoxy)propionsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL545910-25GEA 118 EUR
SIAL545910-25G
3-(4-Fluorphenoxy)propionsäure, Sigma-Aldrich®
3-(4-Fluorphenoxy)propionsäure
Formel: C₉H₉FO₃
MDL Number: MFCD00052838
CAS-Nummer: 1579-78-8
EINECS: 216-424-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR