Benzhydrylamin

Lieferant: Molekula

Synonyme: α-Phenylbenzylamin

MOLEM38567434EA 0 EUR
MOLEM38567434
Benzhydrylamin
Benzhydrylamin
Formel: C₁₃H₁₃N
Molekulargewicht: 183,25 g/mol
MDL Number: MFCD00008059
CAS-Nummer: 91-00-9
EINECS: 202-032-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR