Ethyl-diphenylacetat
Lieferant: LGC Standards PROMOCHEM
PROOMM0025.07EA
1230
EUR
PROOMM0025.07
Ethyl-diphenylacetat
Ethyl-diphenylacetat
Formel:
C₁₆H₁₆O₂ Molekulargewicht: 240,3000 g/mol |
MDL Number:
MFCD00026861 CAS-Nummer: 3468-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...