2-(2-Methylphenyl)ethanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL188123-1GEA 40.9 EUR
SIAL188123-1G SIAL188123-10G
2-(2-Methylphenyl)ethanol, Sigma-Aldrich®
2-(2-Methylphenyl)ethanol
Formel: C₉H₁₂O
MDL Number: MFCD00044241
CAS-Nummer: 19819-98-8
EINECS: 243-349-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR