2-(4-Methylphenyl)ethanol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
SIAL188158-1GEA
44
EUR
SIAL188158-1G
2-(4-Methylphenyl)ethanol, Sigma-Aldrich®
2-(4-Methylphenyl)ethanol
Formel:
C₉H₁₂O Molekulargewicht: 136,20 g/mol |
MDL Number:
MFCD00002904 CAS-Nummer: 699-02-5 EINECS: 211-824-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...