(S)-(+)-Ethyl-3-piperidincarboxylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL696439-1GEA 37.6 EUR
SIAL696439-1G
(S)-(+)-Ethyl-3-piperidincarboxylat, Sigma-Aldrich®
(S)-(+)-Ethyl-3-piperidincarboxylat
Formel: C₈H₁₅NO₂
Molekulargewicht: 157,21 g/mol
MDL Number: MFCD00792499
CAS-Nummer: 37675-18-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR