Jars, with screw cap
Lieferant: Köhler Technische Produkte
216-2953EA 21.8 EUR
216-2953 216-2952 216-2950 216-2956 216-2955 216-2954
Jars, with screw cap
Dosen Dosen, Universal
Jars, made of transparent PS, with screw cap.
  • Temperature resistance: −40 to +75 °C
  • Suitable for the storage of powders and small items