Magnetic stirrer, microwave, Scienceware®
Lieferant: Bel-Art
BELAF370400000EA 617 EUR
BELAF370400000
Magnetic stirrer, microwave, Scienceware®
This microwave stirrer converts the motion from the turntable impeller inside a microwave oven into a magnetic stirrer, which allows simultaneous heating and stirring for increased efficiency.
  • Use with a magnetic stirrer bars up to 5 cm in length
  • Heated stirring can improve dissolving and mixing
  • Permits preparation of super-saturated solutions
Ideal for melting agar and agarose, concentrating urea solutions, or other applications that benefit from heated stirring, and for chemical purification.