Inverses Mikroskop
KRUEMBL3200EA 2599 EUR
KRUEMBL3200
Inverses Mikroskop
This inverted microscope meets the specific needs of biological laboratories.
  • Large working distance to examine samples through Petri dishes
  • Photo and C-mount video adapter for connection of SLR and video cameras
  • Iris diaphragm, green filter and filter holder