SPE plates, 96-well, HyperSep™, Retain PEP
Lieferant: Thermo Scientific
549-0675EA 363 EUR
HYPE60303-208
SPE plates, 96-well, HyperSep™, Retain PEP
Thermo Scientific HyperSep™ Retain™ PEP polymeric solid phase extraction products offers faster throughput and higher reproducibility and recovery in sample preparation. HyperSep™ Retain PEP versatile polymeric SPE material for balanced retention of polar and non-polar analytes. It is packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups.