Capillary columns, TRACE™ TR-225
Lieferant: Thermo Scientific
HYPE260Y142PEA 451 EUR
HYPE260Y142P
Capillary columns, TRACE™ TR-225
These columns are manufactured to minimise the risk of damage from contaminated carrier gas.
  • Mid-polarity phase, 50% cyanopropylphenyl polysiloxane
  • Low bleed even at elevated temperatures
  • Robustness for difficult separations
  • High thermal stability - maximum temperatures up to 250 °C