Ion exchange and size exclusion chromatography column kit, MAb Analysis
Lieferant: Thermo Scientific
HYPE076197EA 1750 EUR
HYPE076197
Ion exchange and size exclusion chromatography column kit, MAb Analysis
The MAb analysis IEX and SEC column kit includes two columns: an ion-exchange (IEX) column and a size exclusion (SEC) column. This kit is a convenient starter and column replacement kit for MAb analysis projects. The kit includes:
MAbPac™ SCX-10 analytical column, 4×250 mm (HYPE074625), a strong cation exchange column designed specifcally for high resolution, high efficiency analysis of monoclonal antibodies and associated variants.
MAbPac™ SEC-1 analytical column, 4×300 mm (HYPE074696), a size exclusion column designed for separating monoclonal antibody (MAb) monomers, aggregates, and fragments.