Digital Manifold
Lieferant: Testo
TEST0563.5702EA 685 EUR
TEST0563.5702
Digital Manifold
  • 'EasyKool' software with extended features
  • Valve block with four connections, four hose parkers and sight glass
  • Integrated vacuum measurement supports evacuation of the system
  • Additional calculation of superheating and subcooling
  • Internal data memory and three temperature probe connections
W×D×H: 280×135×75 mm
Weight: 1200 g
Lieferumfang: Delivery information: Supplied with a calibration protocol, two clamp probes, a transport case and batteries.
Lieferumfang: Supplied with a calibration protocol, two clamp probes, a transport case and batteries.