Waste disposal boxes, Multi-Safe twin
Lieferant: Semadeni
Multi-Safe
SEMA5478EA 10.9 EUR
SEMA5478
Waste disposal boxes, Multi-Safe twin
Behälter Abfallbehälter für spitze/scharfe Abfälle
This safety box equipped with a special introduction slot for the removal of cannulae, scalpels, Butterfly systems, pipes, measurement pipette, blood sedimentation tube, in use syringes, broken glass and long-stemmed objects.
  • Container with conical taper
  • The size of the disposal opening can be further enlarged by breaking out sections as requiered (exeption Multi-Safe twin 700)
  • Lateral carrying handles
  • Hinged day lid
  • Lateral grooves on the lid confirm closure visually
The insertion slot can be enlarged gradually removing the flaps; it can be used in many cases. Universal removal device / torsion suitable for all common types of cannulas, even for insulin injections. All boxes can be secured against overturning by means of suction adapters, or adhesives, or of a support wall.