Single wafer tray, H 20 series, 4" (100 mm)
Lieferant: Entegris
ENTEH203000021216EA 0 EUR
ENTEH203000021216 ENTEH203000011415
Single wafer tray, H 20 series, 4" (100 mm)
Wafer-Handling-Zubehör Wafer-Einsätze
Wafer handling trays to safely transport and protect silicon wafers.
  • For internal handling of single wafers
  • Conical pocket holds one wafer facedown contacting only the wafer’s edges
  • Tweezer slots on tray bottom ease wafer removal
  • Stackable for efficient use of storage space, either tray on tray or cover on tray