Soil extractant solution, Mehlich 2
Lieferant: Hach Lange
HACH2266349EA 0 EUR
HACH2266349
Soil extractant solution, Mehlich 2
Umwelt-Prüfkits
The Mehlich 2 extractant is used for phosphate and potassium determination in soils.