Anti-HNRNPC Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP10728BEA 348 EUR
ABGEAP10728B
Anti-HNRNPC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HNRNPC (heterogeneous nuclear ribonucleoprotein C (C1/C2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR