Anti-DEN1A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9919BEA 348 EUR
ABGEAP9919B
Anti-DEN1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DEN1A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR