Anti-PLA2G6 Rabbit Polyclonal Antibody [clone: 422CT4.6.4]

Lieferant: Abgent
ABGEAP9938CEA 350 EUR
ABGEAP9938C
Anti-PLA2G6 Rabbit Polyclonal Antibody [clone: 422CT4.6.4]
Antikörper
Type: Primary
Antigen: PLA2G6 (phospholipase A2, group VI (cytosolic, calcium-independent))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR