Anti-SNX27 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9947CEA 385 EUR
ABGEAP9947C
Anti-SNX27 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SNX27 (sorting nexin family member 27)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR