Anti-CD300LF Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9948AEA 325 EUR
ABGEAP9948A
Anti-CD300LF Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CD300LF (CD300 molecule-like family member f)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR