Anti-CIRH1A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9951AEA 348 EUR
ABGEAP9951A
Anti-CIRH1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CIRH1A (cirrhosis, autosomal recessive 1A (cirhin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR