Anti-RERE Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9954AEA 360 EUR
ABGEAP9954A
Anti-RERE Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RERE (arginine-glutamic acid dipeptide (RE) repeats)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR