Anti-TAF2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9965AEA 325 EUR
ABGEAP9965A
Anti-TAF2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TAF2 (TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 150kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR