Anti-ELOVL5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9968AEA 348 EUR
ABGEAP9968A
Anti-ELOVL5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ELOVL5 (ELOVL fatty acid elongase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR