Anti-BASP1 Rabbit Polyclonal Antibody [clone: 432CT31.4.2]

Lieferant: Abgent
ABGEAP9970AEA 350 EUR
ABGEAP9970A
Anti-BASP1 Rabbit Polyclonal Antibody [clone: 432CT31.4.2]
Antikörper
Type: Primary
Antigen: BASP1 (brain abundant, membrane attached signal protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR