Anti-GALNT5 Rabbit Polyclonal Antibody [clone: 381CT18.5.1]

Lieferant: Abgent
ABGEAP9976AEA 360 EUR
ABGEAP9976A
Anti-GALNT5 Rabbit Polyclonal Antibody [clone: 381CT18.5.1]
Antikörper
Type: Primary
Antigen: GALNT5 (UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 5 (GalNAc-T5))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR