Anti-ID4 Rabbit Polyclonal Antibody [clone: 384CT24.1.3.4]

Lieferant: Abgent
ABGEAP9977AEA 350 EUR
ABGEAP9977A
Anti-ID4 Rabbit Polyclonal Antibody [clone: 384CT24.1.3.4]
Antikörper
Type: Primary
Antigen: ID4 (inhibitor of DNA binding 4, dominant negative helix-loop-helix protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR