Anti-RHBG Rabbit Polyclonal Antibody [clone: 399CT9.3.4]

Lieferant: Abgent
ABGEAP9979AEA 325 EUR
ABGEAP9979A
Anti-RHBG Rabbit Polyclonal Antibody [clone: 399CT9.3.4]
Antikörper
Type: Primary
Antigen: RHBG (Rh family, B glycoprotein (gene/pseudogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR