Anti-IMMP2L Rabbit Polyclonal Antibody [clone: 429CT16.3.1]

Lieferant: Abgent
ABGEAP9993AEA 350 EUR
ABGEAP9993A
Anti-IMMP2L Rabbit Polyclonal Antibody [clone: 429CT16.3.1]
Antikörper
Type: Primary
Antigen: IMP2L
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR