Anti-ABCC2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1005AEA 350 EUR
ABGEAT1005A
Anti-ABCC2 Mouse Monoclonal Antibody
Type: Primary
Antigen: ABCC2 (ATP-binding cassette, sub-family C (CFTR/MRP), member 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR