Anti-ABCG1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1011AEA 350 EUR
ABGEAT1011A
Anti-ABCG1 Mouse Monoclonal Antibody
Type: Primary
Antigen: ABCG1 (ATP-binding cassette, sub-family G (WHITE), member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR