Anti-ABTB1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1015AEA 350 EUR
ABGEAT1015A
Anti-ABTB1 Mouse Monoclonal Antibody
Type: Primary
Antigen: ABTB1 (ankyrin repeat and BTB (POZ) domain containing 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR