Anti-ACOX2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1027AEA 350 EUR
ABGEAT1027A
Anti-ACOX2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ACOX2 (acyl-CoA oxidase 2, branched chain)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR