Anti-ASNA1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1215AEA 350 EUR
ABGEAT1215A
Anti-ASNA1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ASNA1 (arsA arsenite transporter, ATP-binding, homolog 1 (bacterial))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR