Anti-B4GALT4 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1256AEA 350 EUR
ABGEAT1256A
Anti-B4GALT4 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: B4GALT4 (UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR