Anti-BAAT Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1257AEA 350 EUR
ABGEAT1257A
Anti-BAAT Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: BAAT (bile acid CoA: amino acid N-acyltransferase (glycine N-choloyltransferase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR