Anti-CAP1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1385AEA 350 EUR
ABGEAT1385A
Anti-CAP1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CAP1 (CAP, adenylate cyclase-associated protein 1 (yeast))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR