Anti-CCRN4L Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1426AEA 350 EUR
ABGEAT1426A
Anti-CCRN4L Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CCRN4L (CCR4 carbon catabolite repression 4-like (S. cerevisiae))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR