Anti-CCT6B Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1429AEA 350 EUR
ABGEAT1429A
Anti-CCT6B Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CCT6B (chaperonin containing TCP1, subunit 6B (zeta 2))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR