Anti-CCT8L2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1500AEA 350 EUR
ABGEAT1500A
Anti-CCT8L2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CCT8L2 (chaperonin containing TCP1, subunit 8 (theta)-like 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR