Anti-CKS2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1547AEA 350 EUR
ABGEAT1547A
Anti-CKS2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CKS2 (CDC28 protein kinase regulatory subunit 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR