Anti-COPS2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1591AEA 350 EUR
ABGEAT1591A
Anti-COPS2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: COPS2 (COP9 signalosome subunit 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR