Anti-CRADD Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1616AEA 350 EUR
ABGEAT1616A
Anti-CRADD Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CRADD (CASP2 and RIPK1 domain containing adaptor with death domain)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR