Anti-CSRP3 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1659AEA 350 EUR
ABGEAT1659A
Anti-CSRP3 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CSRP3 (cysteine and glycine-rich protein 3 (cardiac LIM protein))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR